Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMEPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001255976.1 | NP_001242905.1 | 4 | 950 | ||
NM_020182.4 | NP_064567.2 | 4 | 1221 | |||
NM_199169.2 | NP_954638.1 | 4 | 874 | |||
NM_199170.2 | NP_954639.1 | 4 | 867 | |||
NM_199171.2 | NP_954640.1 | 4 | 932 | |||
GenBank mRNA | AF224278.1 | - | 4 | 818 | ||
AF305616.1 | - | 4 | 1148 | |||
AK296199.1 | - | 4 | 874 | |||
AY128643.1 | - | 4 | 822 | |||
BC015918.1 | - | 4 | 808 | |||
BC080635.1 | - | 4 | 807 | 6569922 |