Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGMA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166283.1 | NP_001159755.1 | 2 | 297 | ||
NM_001166286.1 | NP_001159758.1 | 1 | 157 | |||
NM_001166287.1 | NP_001159759.1 | 2 | 271 | |||
NM_001166288.1 | NP_001159760.1 | 2 | 155 | |||
NM_001166289.1 | NP_001159761.1 | 2 | 175 | |||
NM_020211.2 | NP_064596.2 | 2 | 324 | |||
GenBank mRNA | AK074910.1 | - | 2 | 314 | ||
AK074966.1 | - | 1 | 131 | |||
AK074980.1 | - | 2 | 91 | |||
AK125204.1 | - | 2 | 125 | |||
AK299363.1 | - | 2 | 173 | |||
AK309814.1 | - | 2 | 109 | |||
AL136826.1 | - | 2 | 138 | |||
AL390083.1 | - | 1 | 80 | |||
BC015886.1 | - | 1 | 148 | |||
BC140838.1 | - | 2 | 102 | 9021706 | ||
BC151132.1 | - | 2 | 102 | 9057032 | ||
DA183348.1 | - | 2 | 271 | |||
DA295545.1 | - | 2 | 173 | |||
DA453869.1 | - | 1 | 160 | |||
DA512691.1 | - | 2 | 155 | |||
DA805034.1 | - | 2 | 324 | |||
DC317516.1 | - | 2 | 298 |