Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PYY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004160.5 | NP_004151.3 | 6 | 755 | ||
XM_011525035.1 | XP_011523337.1 | 6 | 1365 | |||
GenBank mRNA | D13902.1 | - | 3 | 296 |