Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020232.4 | NP_064617.2 | 5, 6 | 697 | ||
NM_147163.1 | NP_671692.1 | 5, 6 | 555 | |||
GenBank mRNA | AF068295.1 | - | 5, 6 | 555 | ||
AF168712.1 | - | 5, 6 | 629 | |||
AF276707.1 | - | 5, 6 | 1257 | |||
AK025410.1 | - | 1, 2 | 43 | |||
AK057005.1 | - | 5, 6 | 629 | |||
AK225882.1 | - | 5, 6 | 664 | |||
BC013356.2 | - | 5, 6 | 620 | 3640209 |