Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020242.2 | NP_064627.1 | 26 | 3266 | ||
XM_006713264.3 | XP_006713327.1 | 17 | 2124 | |||
XM_011533964.2 | XP_011532266.1 | 25 | 3071 | |||
XM_017006884.1 | XP_016862373.1 | 27 | 3338 | |||
XM_017006885.1 | XP_016862374.1 | 26 | 3188 | |||
XM_017006886.1 | XP_016862375.1 | 28 | 3386 | |||
XM_017006887.1 | XP_016862376.1 | 27 | 3338 | |||
XM_017006888.1 | XP_016862377.1 | 24 | 3005 | |||
XM_017006889.1 | XP_016862378.1 | 27 | 3338 | |||
XM_017006890.1 | XP_016862379.1 | 18 | 2193 | |||
GenBank mRNA | AB035898.1 | - | 26 | 3190 | ||
AF308294.1 | - | 8 | 763 | |||
AK027816.1 | - | 8 | 872 | |||
AK307674.1 | - | 26 | 3185 | |||
AL832908.1 | - | 17 | 2105 | |||
BC117174.1 | - | 25 | 3119 | 40125725 | ||
BC143752.1 | - | 25 | 3120 |