Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020242.2 | NP_064627.1 | 30 | 3813 | ||
XM_006713264.3 | XP_006713327.1 | 21 | 2671 | |||
XM_011533964.2 | XP_011532266.1 | 29 | 3618 | |||
XM_017006884.1 | XP_016862373.1 | 31 | 3885 | |||
XM_017006885.1 | XP_016862374.1 | 30 | 3735 | |||
XM_017006886.1 | XP_016862375.1 | 32 | 3933 | |||
XM_017006887.1 | XP_016862376.1 | 31 | 3885 | |||
XM_017006888.1 | XP_016862377.1 | 28 | 3552 | |||
XM_017006890.1 | XP_016862379.1 | 22 | 2740 | |||
GenBank mRNA | AB035898.1 | - | 30 | 3737 | ||
AF308294.1 | - | 12 | 1306 | |||
AK027816.1 | - | 12 | 1419 | |||
AL832908.1 | - | 21 | 2652 | |||
BC117174.1 | - | 29 | 3666 | 40125725 | ||
BC143752.1 | - | 29 | 3667 |