Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COQ8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020247.4 | NP_064632.2 | 10, 11 | 1426 | ||
XM_005273201.1 | XP_005273258.1 | 10, 11 | 1322 | |||
XM_011544238.1 | XP_011542540.1 | 11, 12 | 1518 | |||
XM_011544239.2 | XP_011542541.1 | 12, 13 | 1541 | |||
XM_011544240.2 | XP_011542542.1 | 10, 11 | 1335 | |||
XM_011544241.2 | XP_011542543.1 | 10, 11 | 1297 | |||
XM_017001852.1 | XP_016857341.1 | 10, 11 | 1516 | |||
GenBank mRNA | AB073905.1 | - | 10, 11 | 1349 | ||
AJ278126.1 | - | 6, 7 | 644 | |||
AK074693.1 | - | 10, 11 | 1369 | |||
AK090494.1 | - | 10, 11 | 1163 | |||
AK092784.1 | - | 10, 11 | 1149 | |||
AK297735.1 | - | 7, 8 | 705 | |||
AK301012.1 | - | 12, 13 | 1347 | |||
AL137511.1 | - | 5, 6 | 1410 | |||
BC005171.2 | - | 10, 11 | 1375 | 3604948 | ||
BX648860.1 | - | 10, 11 | 1247 |