Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COQ9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020312.3 | NP_064708.1 | 4 | 507 | ||
GenBank mRNA | AK290627.1 | - | 4 | 437 | ||
AK293585.1 | - | 4 | 441 | |||
AK295793.1 | - | 4 | 450 | |||
AK303527.1 | - | 4 | 430 | |||
BC001478.2 | - | 2 | 82 | |||
BC029341.1 | - | 4 | 441 | |||
BC054340.1 | - | 4 | 441 | 6514554 | ||
BC064946.1 | - | 4 | 442 | 6023461 |