Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIAPIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308347.1 | NP_001295276.1 | 2 | 280 | ||
NM_001308358.1 | NP_001295287.1 | 2 | 280 | |||
NM_020313.3 | NP_064709.2 | 2 | 280 | |||
XM_005256063.4 | XP_005256120.1 | 2 | 171 | |||
GenBank mRNA | AF248964.1 | - | 2 | 184 | ||
AK074263.1 | - | 2 | 121 | |||
AK292281.1 | - | 2 | 123 | |||
AK295024.1 | - | 1 | 513 | |||
AL136613.1 | - | 2 | 141 | |||
BC002568.2 | - | 2 | 122 | 3139226 | ||
BC024196.2 | - | 2 | 161 | 4816594 | ||
BC067303.1 | - | 2 | 280 | 30389410 | ||
BC071740.1 | - | 2 | 155 | 5563094 | ||
BG746807.1 | - | 1 | 126 | |||
BJ991015.1 | - | 2 | 161 | |||
CR533545.1 | - | 1 | 38 |