Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174100.1 | NP_001167571.1 | 14 | 1509 | ||
NM_020418.3 | NP_065151.2 | 13 | 1283 | |||
NM_033008.2 | NP_127501.1 | 13 | 1361 | |||
NM_033010.2 | NP_127503.1 | 13 | 1303 | |||
XM_005265330.2 | XP_005265387.1 | 14 | 3316 | |||
XM_005265331.2 | XP_005265388.1 | 14 | 1435 | |||
XM_005265332.2 | XP_005265389.1 | 13 | 3187 | |||
XM_006713268.2 | XP_006713331.1 | 14 | 3124 | |||
XM_006713269.1 | XP_006713332.1 | 15 | 1580 | |||
XM_006713270.1 | XP_006713333.1 | 12 | 1223 | |||
XM_006713271.2 | XP_006713334.1 | 14 | 3119 | |||
XM_017006916.1 | XP_016862405.1 | 15 | 1737 | |||
XM_017006917.1 | XP_016862406.1 | 14 | 2927 | |||
XM_017006918.1 | XP_016862407.1 | 13 | 2779 | |||
XM_017006919.1 | XP_016862408.1 | 13 | 1155 | |||
XM_017006920.1 | XP_016862409.1 | 12 | 1026 | |||
GenBank mRNA | AF141340.1 | - | 7 | 685 | ||
AF176330.1 | - | 13 | 1303 | |||
AF257770.1 | - | 10 | 1940 | |||
AF257771.1 | - | 11 | 1775 | |||
AK001244.1 | - | 11 | 1430 | |||
AK023993.1 | - | 13 | 1350 | |||
AK225815.1 | - | 14 | 1487 | |||
BC003008.1 | - | 13 | 1324 | 2822683 | ||
BC004153.2 | - | 13 | 1324 | 2822683 | ||
BC017098.1 | - | 13 | 1278 | 3508303 | ||
BU684420.1 | - | 1 | 676 | |||
BX647811.1 | - | 12 | 1890 |