Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191037.1 | NP_001177966.1 | 11 | 1386 | ||
NM_002807.3 | NP_002798.2 | 11 | 1386 | |||
NR_034059.1 | 10 | 1216 | ||||
XM_017004517.1 | XP_016860006.1 | 11 | 1386 | |||
GenBank mRNA | AK307415.1 | - | 10 | 1213 | ||
AK307513.1 | - | 9 | 1088 | |||
AK312528.1 | - | 11 | 1343 | |||
BC001053.2 | - | 11 | 1343 | |||
BC014013.2 | - | 11 | 1355 | |||
BC020223.1 | - | 11 | 1333 | |||
BC032445.1 | - | 11 | 1358 | |||
BC039845.1 | - | 11 | 1309 | |||
BC047897.1 | - | 11 | 1308 | |||
BC064398.1 | - | 11 | 1357 | |||
BC073833.1 | - | 11 | 1310 | |||
BC094720.1 | - | 11 | 1372 | 4826340 | ||
BC112344.1 | - | 11 | 1347 | 40008454 | ||
BC114434.1 | - | 11 | 1347 | 40008444 | ||
D44466.1 | - | 11 | 1304 | |||
KU921434.1 | - | 4 | 343 |