Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KNL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144508.4 | NP_653091.3 | 10 | 1020 | ||
NM_170589.4 | NP_733468.3 | 11 | 1098 | |||
XM_017022432.1 | XP_016877921.1 | 9 | 1223 | |||
GenBank mRNA | AB022190.1 | - | 10 | 1021 | ||
AF173994.1 | - | 11 | 1327 | |||
AF248041.1 | - | 10 | 1004 | |||
AF461041.1 | - | 10 | 1004 | |||
AK307605.1 | - | 10 | 1012 | |||
AK307996.1 | - | 10 | 1010 | |||
BC029373.1 | - | 10 | 997 |