Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC17A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020346.2 | NP_065079.1 | 12 | 2043 | ||
GenBank mRNA | AB032435.1 | - | 12 | 2043 | ||
AK294502.1 | - | 11 | 1413 | |||
AK309131.1 | - | 10 | 1296 | |||
BC069629.1 | - | 12 | 1789 | 7262471 | ||
BC069640.1 | - | 12 | 1790 | 7262388 | ||
BC069646.1 | - | 12 | 1790 | 7262400 |