Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCNP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320397.1 | NP_001307326.1 | 4, 5 | 429 | ||
NM_001320398.1 | NP_001307327.1 | 4, 5 | 525 | |||
NM_001320399.1 | NP_001307328.1 | 5, 6 | 654 | |||
NM_001320401.1 | NP_001307330.1 | 3, 4 | 453 | |||
NM_020357.2 | NP_065090.1 | 4, 5 | 528 | |||
NR_135223.1 | 4, 5 | 476 | ||||
NR_135224.1 | 3, 4 | 313 | ||||
NR_135225.1 | 4, 5 | 473 | ||||
NR_135227.1 | 4, 5 | 729 | ||||
NR_135229.1 | 4, 5 | 657 | ||||
NR_135230.1 | 3, 4 | 401 | ||||
NR_135231.1 | 2, 3 | 238 | ||||
NR_135232.1 | 5, 6 | 571 | ||||
GenBank mRNA | AB037675.1 | - | 4, 5 | 424 | ||
AK025551.1 | - | 4, 5 | 368 | |||
AK098760.1 | - | 4, 5 | 408 | |||
AK222978.1 | - | 5, 6 | 419 | |||
AK314629.1 | - | 4, 5 | 409 | |||
BC022001.2 | - | 4, 5 | 426 | 4823990 | ||
BC107796.1 | - | 4, 5 | 370 | |||
BI596913.1 | - | 5, 6 | 580 | |||
CN482876.1 | - | 5, 6 | 455 | |||
CR627371.1 | - | 3, 4 | 214 | |||
DA304502.1 | - | 4, 5 | 476 | |||
DA710370.1 | - | 3, 4 | 343 | |||
DA817951.1 | - | 4, 5 | 393 | |||
DB032930.1 | - | 3, 4 | 252 | |||
DB060329.1 | - | 4, 5 | 340 | |||
DB481581.1 | - | 3, 4 | 289 | |||
HY051831.1 | - | 2, 3 | 126 |