Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNPLA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020376.3 | NP_065109.1 | 3 | 586 | ||
XM_006718265.3 | XP_006718328.1 | 3 | 592 | |||
XM_006718266.3 | XP_006718329.1 | 2 | 532 | |||
XM_017018028.1 | XP_016873517.1 | 2 | 532 | |||
GenBank mRNA | AF055000.1 | - | 1 | 104 | ||
AJ278475.1 | - | 3 | 455 | |||
AJ278476.1 | - | 2 | 475 | |||
AK308435.1 | - | 2 | 552 | |||
AY203925.1 | - | 1 | 990 | |||
AY894804.1 | - | 2 | 383 | |||
BC017280.2 | - | 3 | 585 | 4875483 | ||
HQ651812.1 | - | 3 | 586 | |||
JF279441.1 | - | 3 | 586 | |||
JF279442.1 | - | 3 | 586 | |||
KF601369.1 | - | 3 | 586 | |||
KF833264.1 | - | 2 | 383 | |||
KF833265.1 | - | 2 | 383 |