Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF695 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020394.4 | NP_065127.4 | 4 | 563 | ||
NR_037892.1 | 4 | 563 | ||||
GenBank mRNA | BC023527.2 | - | 4 | 471 | ||
BE743952.1 | - | 3 | 302 | |||
CN394881.1 | - | 4 | 495 |
Gene Symbol: ZNF670-ZNF695 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037894.1 | 4 | 559 | |||
GenBank mRNA | AK126366.1 | - | 6 | 718 | ||
AL524527.3 | - | 4 | 482 |