Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPPIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302861.1 | NP_001289790.1 | 2 | 189 | ||
NM_020398.3 | NP_065131.1 | 2 | 189 | |||
GenBank mRNA | AF286368.1 | - | 2 | 141 | ||
AF286369.1 | - | 1 | 636 | |||
AF286370.2 | - | 2 | 141 | |||
AK126022.1 | - | 2 | 120 | |||
AK311906.1 | - | 2 | 188 | |||
AL118493.1 | - | 2 | 143 | |||
BC003675.1 | - | 2 | 164 | |||
BC044829.1 | - | 2 | 152 | 5171493 | ||
BC053369.1 | - | 2 | 147 | 5165509 | ||
DA821702.1 | - | 2 | 164 | |||
DC395362.1 | - | 2 | 190 |
Gene Symbol: EPPIN-WFDC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198986.1 | NP_001185915.1 | 2 | 189 | ||
GenBank mRNA | AK301937.1 | - | 2 | 164 |