Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMD13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002817.3 | NP_002808.3 | 4 | 479 | ||
NM_175932.2 | NP_787128.2 | 2 | 485 | |||
XM_011520235.2 | XP_011518537.1 | 4 | 612 | |||
GenBank mRNA | AB009398.1 | - | 4 | 306 | ||
AK094775.1 | - | 2 | 316 | |||
AK222895.1 | - | 4 | 310 | |||
AK314733.1 | - | 4 | 741 |