Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSME2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002818.2 | NP_002809.2 | 4, 5 | 294 | ||
XM_006720213.1 | XP_006720276.1 | 4, 5 | 1088 | |||
GenBank mRNA | AK026580.1 | - | 4, 5 | 315 | ||
AK225876.1 | - | 4, 5 | 257 | |||
AK311569.1 | - | 2, 3 | 933 | |||
AY771595.1 | - | 4, 5 | 243 | |||
BC004368.1 | - | 4, 5 | 244 | 3640584 | ||
BC019885.1 | - | 4, 5 | 244 | 4994572 | ||
BC072025.1 | - | 4, 5 | 243 | 6270762 | ||
BX161498.1 | - | 4, 5 | 869 | |||
CR541657.1 | - | 4, 5 | 229 | |||
CR541743.1 | - | 4, 5 | 229 | |||
D45248.1 | - | 4, 5 | 294 |