Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSPH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004577.3 | NP_004568.2 | 7, 8 | 1288 | ||
XM_005271773.1 | XP_005271830.1 | 7, 8 | 1358 | |||
XM_005271774.1 | XP_005271831.1 | 8, 9 | 1474 | |||
XM_005271775.1 | XP_005271832.1 | 7, 8 | 1314 | |||
XM_005271776.1 | XP_005271833.1 | 8, 9 | 1422 | |||
XM_006715760.1 | XP_006715823.1 | 8, 9 | 1472 | |||
XM_006715761.1 | XP_006715824.1 | 6, 7 | 1223 | |||
XM_011515461.1 | XP_011513763.1 | 8, 9 | 1428 | |||
XM_017012466.1 | XP_016867955.1 | 9, 10 | 1588 | |||
XM_017012467.1 | XP_016867956.1 | 9, 10 | 1684 | |||
XM_017012468.1 | XP_016867957.1 | 7, 8 | 1337 | |||
XM_017012469.1 | XP_016867958.1 | 7, 8 | 1287 | |||
GenBank mRNA | AK223508.1 | - | 4, 5 | 578 | ||
AK315235.1 | - | 6, 7 | 900 | |||
BC063614.1 | - | 7, 8 | 1136 | 3850060 | ||
BX537439.1 | - | 7, 8 | 1258 |