Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020650.2 | NP_065701.2 | 7 | 1384 | ||
XM_005259089.3 | XP_005259146.1 | 8 | 1479 | |||
XM_011527143.1 | XP_011525445.1 | 8 | 1573 | |||
XM_017027023.1 | XP_016882512.1 | 7 | 1658 | |||
GenBank mRNA | AF183423.1 | - | 6 | 969 | ||
AY195859.1 | - | 7 | 1384 | |||
AY358423.1 | - | 7 | 971 | |||
BC013436.1 | - | 7 | 967 | 3889718 |