Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF286A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130842.1 | NP_001124314.1 | 2 | 414 | ||
NM_001288642.1 | NP_001275571.1 | 2 | 341 | |||
NM_001288645.1 | NP_001275574.1 | 2 | 501 | |||
NM_001288646.1 | NP_001275575.1 | 2 | 414 | |||
NM_001288647.1 | NP_001275576.1 | 2 | 414 | |||
NM_020652.2 | NP_065703.1 | 2 | 501 | |||
GenBank mRNA | AF217226.1 | - | 1 | 333 | ||
AK092259.1 | - | 2 | 339 | |||
AK315062.1 | - | 2 | 259 | |||
BC015730.1 | - | 2 | 339 | |||
BC025260.1 | - | 2 | 339 | |||
BC128259.1 | - | 1 | 169 | 40116891 | ||
BC128411.1 | - | 1 | 169 | 40116881 | ||
BP193773.1 | - | 2 | 414 | |||
DA188283.1 | - | 2 | 272 | |||
DA748526.1 | - | 2 | 252 | |||
DN997307.1 | - | 1 | 197 |
Gene Symbol: ZNF286B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145045.1 | NP_001138517.1 | 2 | 269 |