Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF286A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130842.1 | NP_001124314.1 | 5 | 693 | ||
NM_001288642.1 | NP_001275571.1 | 5 | 620 | |||
NM_001288643.1 | NP_001275572.1 | 4 | 461 | |||
NM_001288644.1 | NP_001275573.1 | 3 | 346 | |||
NM_001288645.1 | NP_001275574.1 | 5 | 780 | |||
NM_001288646.1 | NP_001275575.1 | 5 | 693 | |||
NM_001288647.1 | NP_001275576.1 | 4 | 578 | |||
NM_001288648.1 | NP_001275577.1 | 4 | 461 | |||
NM_001288649.1 | NP_001275578.1 | 3 | 282 | |||
NM_020652.2 | NP_065703.1 | 5 | 780 | |||
GenBank mRNA | AF086305.1 | - | 3 | 213 | ||
AF217226.1 | - | 4 | 612 | |||
AK092259.1 | - | 5 | 618 | |||
AK315062.1 | - | 5 | 538 | |||
BC015730.1 | - | 5 | 618 | |||
BC025260.1 | - | 5 | 618 | |||
BC128259.1 | - | 4 | 448 | 40116891 | ||
BC128411.1 | - | 4 | 448 | 40116881 | ||
DA096356.1 | - | 4 | 284 | |||
DA188283.1 | - | 4 | 436 | |||
DA748526.1 | - | 5 | 531 | |||
DA786711.1 | - | 3 | 120 | |||
DA796080.1 | - | 5 | 453 | |||
DA806764.1 | - | 3 | 194 | |||
DC344287.1 | - | 4 | 347 | |||
DN997307.1 | - | 4 | 476 |