Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S100A14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020672.2 | NP_065723.1 | 2, 3 | 146 | ||
XM_005245362.1 | XP_005245419.1 | 2, 3 | 142 | |||
XM_017001875.1 | XP_016857364.1 | 3, 4 | 234 | |||
GenBank mRNA | AY007220.1 | - | 2, 3 | 128 | ||
BC005019.1 | - | 1, 2 | 110 | 3640899 | ||
BG677973.1 | - | 2, 3 | 146 |