Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIF4GD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242498.1 | NP_001229427.1 | 4, 5 | 450 | ||
NM_001242500.1 | NP_001229429.1 | 3, 4 | 352 | |||
NM_001242501.1 | NP_001229430.1 | 2, 3 | 250 | |||
NM_020679.3 | NP_065730.2 | 4, 5 | 429 | |||
XM_005257530.1 | XP_005257587.1 | 4, 5 | 571 | |||
XM_005257532.4 | XP_005257589.1 | 3, 4 | 327 | |||
XM_005257533.1 | XP_005257590.1 | 3, 4 | 426 | |||
XM_011525051.2 | XP_011523353.1 | 4, 5 | 666 | |||
XM_011525052.2 | XP_011523354.1 | 4, 5 | 645 | |||
XM_011525053.2 | XP_011523355.1 | 3, 4 | 543 | |||
XM_017024888.1 | XP_016880377.1 | 4, 5 | 666 | |||
XM_017024889.1 | XP_016880378.1 | 4, 5 | 450 | |||
XM_017024890.1 | XP_016880379.1 | 4, 5 | 476 | |||
XM_017024891.1 | XP_016880380.1 | 4, 5 | 388 | |||
XM_017024893.1 | XP_016880382.1 | 3, 4 | 822 | |||
XM_017024894.1 | XP_016880383.1 | 3, 4 | 457 | |||
GenBank mRNA | AF225422.1 | - | 2, 3 | 446 | ||
AK300711.1 | - | 4, 5 | 389 | |||
BC033759.1 | - | 3, 4 | 328 | 5215495 | ||
BI752882.1 | - | 4, 5 | 447 | |||
BU190758.1 | - | 3, 4 | 351 | |||
CA488406.1 | - | 2, 3 | 250 | |||
KU178731.1 | - | 3, 4 | 292 | |||
KU178732.1 | - | 3, 4 | 313 | |||
KU178733.1 | - | 2, 3 | 190 |