Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCYL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048218.1 | NP_001041683.1 | 10 | 1352 | ||
NM_020680.3 | NP_065731.3 | 10 | 1352 | |||
XM_005274118.3 | XP_005274175.1 | 10 | 1364 | |||
XM_005274120.3 | XP_005274177.1 | 10 | 1364 | |||
XM_005274121.3 | XP_005274178.1 | 10 | 1364 | |||
XM_017018029.1 | XP_016873518.1 | 10 | 1364 | |||
XM_017018030.1 | XP_016873519.1 | 10 | 1364 | |||
XM_017018031.1 | XP_016873520.1 | 10 | 1364 | |||
XM_017018032.1 | XP_016873521.1 | 10 | 1206 | |||
XM_017018033.1 | XP_016873522.1 | 10 | 1356 | |||
GenBank mRNA | AB047077.1 | - | 10 | 1275 | ||
AB051427.1 | - | 10 | 1275 | |||
AB051428.1 | - | 10 | 1275 | |||
AF225424.1 | - | 4 | 416 | |||
AF297709.1 | - | 10 | 1314 | |||
AK314945.1 | - | 10 | 1308 | |||
BC009967.2 | - | 4 | 350 | |||
BC069233.1 | - | 10 | 1316 | 6581110 |