Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024100.3 | NP_077005.2 | 1 | 89 | ||
XM_017027027.1 | XP_016882516.1 | 3 | 615 | |||
GenBank mRNA | AL560525.3 | - | 1 | 59 | ||
BC000040.2 | - | 1 | 74 | 3507673 | ||
BC001648.2 | - | 1 | 69 | 2819452 | ||
CB991338.1 | - | 116 |