Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNT16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168368.1 | NP_001161840.1 | 14 | 1752 | ||
NM_020692.2 | NP_065743.2 | 14 | 1752 | |||
XM_011537005.1 | XP_011535307.1 | 14 | 1564 | |||
XM_011537006.2 | XP_011535308.1 | 14 | 1564 | |||
XM_011537007.2 | XP_011535309.1 | 14 | 1564 | |||
XM_017021498.1 | XP_016876987.1 | 15 | 1739 | |||
GenBank mRNA | AB032956.1 | - | 14 | 1542 | ||
AB078143.1 | - | 14 | 1425 | |||
AJ505951.1 | - | 14 | 1425 | |||
AK289745.1 | - | 14 | 1511 | |||
AK299588.1 | - | 5 | 455 | |||
AK315848.1 | - | 14 | 1516 | |||
BC036812.1 | - | 14 | 1503 | 5734009 | ||
BC098578.1 | - | 14 | 1518 | 30924466 | ||
BC122545.1 | - | 14 | 1525 | 40084363 |