Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020705.2 | NP_065756.1 | 2, 3 | 1101 | ||
XM_017023494.1 | XP_016878983.1 | 2, 3 | 1231 | |||
XM_017023495.1 | XP_016878984.1 | 2, 3 | 1143 | |||
GenBank mRNA | BC112389.1 | - | 2, 3 | 1101 | 5502314 | |
BC127014.1 | - | 1, 2 | 1015 | 40118225 |