Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCAF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145444.1 | NP_001138916.1 | 2 | 517 | ||
NM_001145445.1 | NP_001138917.1 | 2 | 517 | |||
NM_020706.2 | NP_065757.1 | 2 | 517 | |||
XM_005261017.2 | XP_005261074.1 | 2 | 498 | |||
XM_006724035.2 | XP_006724098.1 | 2 | 498 | |||
XM_006724036.2 | XP_006724099.1 | 2 | 498 | |||
XM_017028415.1 | XP_016883904.1 | 2 | 478 | |||
XM_017028416.1 | XP_016883905.1 | 2 | 363 | |||
XM_017028417.1 | XP_016883906.1 | 2 | 498 | |||
GenBank mRNA | AF023142.1 | - | 2 | 121 | ||
AK057840.1 | - | 2 | 360 | |||
AK308406.1 | - | 2 | 367 | |||
AL834304.1 | - | 2 | 297 | |||
BC014921.1 | - | 2 | 494 | 3907610 | ||
BC043353.1 | - | 2 | 474 | 6019414 | ||
BC052286.1 | - | 2 | 348 | 5442227 | ||
BC064990.1 | - | 2 | 348 | 5432277 |