Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESYT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020728.2 | NP_065779.1 | 17 | 2119 | ||
GenBank mRNA | AB033054.2 | - | 18 | 1905 | ||
AK001181.1 | - | 6 | 608 | |||
AK124091.1 | - | 2 | 867 | |||
AK308824.1 | - | 17 | 1727 | |||
AL833233.1 | - | 17 | 1728 | |||
AY368150.1 | - | 17 | 2119 | |||
BC013957.2 | - | 9 | 841 | |||
DQ993201.1 | - | 19 | 2137 |