Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESYT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020728.2 | NP_065779.1 | 6 | 935 | ||
GenBank mRNA | AB033054.2 | - | 7 | 721 | ||
AK126214.1 | - | 6 | 580 | |||
AK308824.1 | - | 6 | 543 | |||
AL833233.1 | - | 6 | 544 | |||
AY368150.1 | - | 6 | 935 | |||
DQ993201.1 | - | 8 | 953 |