Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIDINS220 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020738.2 | NP_065789.1 | 30 | 5088 | ||
GenBank mRNA | AB033076.2 | - | 30 | 5063 | ||
AK022873.1 | - | 5 | 1341 | |||
AK025528.1 | - | 1 | 756 | |||
AK294464.1 | - | 6 | 1663 | |||
AK295358.1 | - | 5 | 1551 | |||
AL137553.1 | - | 1 | 332 | |||
BC094714.1 | - | 1 | 476 | |||
BC130610.1 | - | 29 | 5012 | 40146641 | ||
BX538189.1 | - | 6 | 1668 | |||
BX640878.1 | - | 6 | 1587 | |||
KJ812119.1 | - | 7 | 1714 | |||
KJ812120.1 | - | 6 | 1657 | |||
KJ812121.1 | - | 5 | 1543 |