Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGAG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020769.2 | NP_065820.1 | 3 | 366 | ||
XM_011530998.2 | XP_011529300.1 | 3 | 388 | |||
XM_011530999.2 | XP_011529301.1 | 4 | 1237 | |||
XM_017029694.1 | XP_016885183.1 | 2 | 296 | |||
XM_017029695.1 | XP_016885184.1 | 5 | 1370 | |||
GenBank mRNA | AB037739.1 | - | 3 | 366 | ||
AK302427.1 | - | 3 | 388 | |||
AY121804.1 | - | 3 | 366 | |||
BC113670.1 | - | 2 | 207 | 8322722 | ||
BC117184.1 | - | 2 | 207 | 40125735 |