Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006657.1 | NP_001006658.1 | 16 | 1760 | ||
NM_020779.3 | NP_065830.2 | 15 | 1727 | |||
XM_011533007.2 | XP_011531309.1 | 5 | 452 | |||
GenBank mRNA | AB037757.2 | - | 16 | 1768 | ||
AK091205.1 | - | 9 | 1095 | |||
AK122917.1 | - | 15 | 1652 | |||
AK315094.1 | - | 15 | 1652 | |||
AL832903.1 | - | 4 | 257 | |||
BC036659.2 | - | 15 | 1726 | 5266940 |