Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006657.1 | NP_001006658.1 | 20 | 2232 | ||
NM_020779.3 | NP_065830.2 | 19 | 2199 | |||
XM_011533007.2 | XP_011531309.1 | 9 | 924 | |||
GenBank mRNA | AB037757.2 | - | 20 | 2240 | ||
AK091205.1 | - | 13 | 1567 | |||
AK122917.1 | - | 19 | 2124 | |||
AK315094.1 | - | 19 | 2124 | |||
AL832903.1 | - | 8 | 729 | |||
BC036659.2 | - | 19 | 2198 | 5266940 |