Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIPA1L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020808.4 | NP_065859.3 | 12 | 3877 | ||
XM_005273211.2 | XP_005273268.1 | 14 | 4214 | |||
XM_005273212.4 | XP_005273269.1 | 14 | 4021 | |||
XM_005273213.4 | XP_005273270.1 | 13 | 3914 | |||
XM_011544243.2 | XP_011542545.1 | 15 | 4070 | |||
XM_017001896.1 | XP_016857385.1 | 14 | 3967 | |||
XM_017001897.1 | XP_016857386.1 | 14 | 4214 | |||
XM_017001898.1 | XP_016857387.1 | 14 | 3988 | |||
GenBank mRNA | AB037810.1 | - | 12 | 3081 | ||
AK022852.1 | - | 4 | 477 | |||
AK093191.1 | - | 6 | 1013 | |||
AY168879.1 | - | 12 | 3837 | |||
BC013119.1 | - | 4 | 388 | |||
BC082251.1 | - | 4 | 417 | |||
BC150334.1 | - | 12 | 3092 | 8860510 |