Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122819.2 | NP_001116291.1 | 4, 5 | 969 | ||
NM_001287212.1 | NP_001274141.1 | 4, 5 | 1096 | |||
NM_020816.3 | NP_065867.2 | 4, 5 | 969 | |||
XM_005245950.3 | XP_005246007.1 | 4, 5 | 722 | |||
XM_005245951.3 | XP_005246008.1 | 4, 5 | 722 | |||
XM_011541837.1 | XP_011540139.1 | 4, 5 | 722 | |||
XM_011541838.1 | XP_011540140.1 | 4, 5 | 722 | |||
XM_011541839.2 | XP_011540141.1 | 4, 5 | 722 | |||
XM_011541840.2 | XP_011540142.1 | 4, 5 | 730 | |||
XM_011541841.2 | XP_011540143.1 | 4, 5 | 722 | |||
XM_011541842.1 | XP_011540144.1 | 4, 5 | 722 | |||
XM_011541843.1 | XP_011540145.1 | 4, 5 | 722 | |||
XM_011541844.2 | XP_011540146.1 | 4, 5 | 722 | |||
XM_011541845.2 | XP_011540147.1 | 4, 5 | 722 | |||
XM_011541846.2 | XP_011540148.1 | 4, 5 | 493 | |||
GenBank mRNA | AB037826.2 | - | 4, 5 | 570 | ||
AB290162.1 | - | 4, 5 | 776 | |||
AY484427.1 | - | 4, 5 | 968 | |||
BC065927.1 | - | 4, 5 | 732 | 6171598 |