Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020827.2 | NP_065878.1 | 5 | 1729 | ||
XM_017008483.1 | XP_016863972.1 | 5 | 1987 | |||
XM_017008484.1 | XP_016863973.1 | 5 | 1621 | |||
XM_017008485.1 | XP_016863974.1 | 7 | 1854 | |||
XM_017008486.1 | XP_016863975.1 | 5 | 1621 | |||
GenBank mRNA | AB037851.1 | - | 4 | 1569 | ||
AK091993.1 | - | 5 | 1660 | |||
AL133615.1 | - | 4 | 697 |