Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128833.1 | NP_001122305.1 | 4 | 1767 | ||
NM_020899.3 | NP_065950.2 | 4 | 1843 | |||
XM_006721563.3 | XP_006721626.1 | 4 | 1694 | |||
XM_006721564.2 | XP_006721627.1 | 4 | 1674 | |||
XM_011523972.2 | XP_011522274.1 | 4 | 2126 | |||
GenBank mRNA | AB040971.2 | - | 4 | 1767 | ||
AK122826.1 | - | 2 | 1095 | |||
AK122971.1 | - | 4 | 1667 | |||
AY302699.1 | - | 4 | 1683 | |||
BC043352.2 | - | 4 | 1792 | 6175382 |