Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002827.3 | NP_002818.1 | 2 | 290 | ||
GenBank mRNA | AK290638.1 | - | 2 | 257 | ||
AK316563.1 | - | 2 | 232 | |||
AU117677.1 | - | 2 | 257 | |||
BC015660.2 | - | 2 | 219 | 4844022 | ||
BC018164.1 | - | 2 | 200 | 4338750 | ||
BT006752.1 | - | 2 | 83 | |||
M31724.1 | - | 2 | 173 | |||
M33689.1 | - | 2 | 155 |