Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPTBN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020971.2 | NP_066022.2 | 26 | 5612 | ||
NM_025213.2 | NP_079489.2 | 9 | 1665 | |||
XM_011527173.2 | XP_011525475.1 | 26 | 5678 | |||
XM_017027049.1 | XP_016882538.1 | 26 | 5865 | |||
XM_017027050.1 | XP_016882539.1 | 24 | 5247 | |||
XM_017027051.1 | XP_016882540.1 | 26 | 5678 | |||
GenBank mRNA | AB046862.1 | - | 17 | 4355 | ||
AF082075.1 | - | 26 | 5673 | |||
AF311855.1 | - | 26 | 5612 | |||
AF311856.1 | - | 9 | 1665 | |||
AF324063.1 | - | 26 | 5596 | |||
AF324064.1 | - | 26 | 5596 | |||
AL133093.1 | - | 4 | 828 | |||
AY004226.1 | - | 26 | 5581 | |||
AY004227.1 | - | 25 | 5495 |