Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPTBN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020971.2 | NP_066022.2 | 20 | 4437 | ||
NM_025213.2 | NP_079489.2 | 3 | 490 | |||
XM_011527173.2 | XP_011525475.1 | 20 | 4503 | |||
XM_017027049.1 | XP_016882538.1 | 20 | 4690 | |||
XM_017027050.1 | XP_016882539.1 | 18 | 4072 | |||
XM_017027051.1 | XP_016882540.1 | 20 | 4503 | |||
GenBank mRNA | AB046862.1 | - | 11 | 3180 | ||
AF082075.1 | - | 20 | 4498 | |||
AF311855.1 | - | 20 | 4437 | |||
AF311856.1 | - | 3 | 490 | |||
AF324063.1 | - | 20 | 4421 | |||
AF324064.1 | - | 20 | 4421 | |||
AK308547.1 | - | 3 | 478 | |||
AK309418.1 | - | 3 | 452 | |||
AY004226.1 | - | 20 | 4406 | |||
AY004227.1 | - | 19 | 4320 | |||
DA514440.1 | - | 3 | 452 |