Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCAR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021174.5 | NP_066997.3 | 13 | 1905 | ||
NR_033902.1 | 13 | 1723 | ||||
XM_011544603.1 | XP_011542905.1 | 13 | 1723 | |||
XM_011544604.1 | XP_011542906.1 | 12 | 1603 | |||
XM_017013717.1 | XP_016869206.1 | 13 | 1905 | |||
GenBank mRNA | AB075847.1 | - | 10 | 1214 | ||
AF293335.1 | - | 2 | 161 | |||
AK095712.1 | - | 13 | 1579 | |||
AK096547.1 | - | 4 | 554 | |||
AK314535.1 | - | 13 | 1598 | |||
AL834162.1 | - | 13 | 1892 | |||
AL834351.1 | - | 13 | 1893 | |||
AL834352.1 | - | 13 | 1893 | |||
BC065495.1 | - | 13 | 1581 | 5496068 | ||
BX640952.1 | - | 13 | 1592 |