Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001188.3 | NP_001179.1 | 4 | 623 | ||
XM_011514779.2 | XP_011513081.1 | 5 | 734 | |||
XM_011514780.1 | XP_011513082.1 | 4 | 544 | |||
GenBank mRNA | AF520590.1 | - | 3 | 260 | ||
AK091807.1 | - | 4 | 488 | |||
AK292093.1 | - | 5 | 661 | |||
AK293547.1 | - | 4 | 553 | |||
AK303422.1 | - | 4 | 602 | |||
BC004431.1 | - | 1 | 534 | 2819507 | ||
BC110337.1 | - | 4 | 533 | 5755200 | ||
CR457419.1 | - | 3 | 323 | |||
CR627020.1 | - | 1 | 4748 | |||
KU177934.1 | - | 3 | 323 | |||
KU177935.1 | - | 3 | 323 | |||
U16811.1 | - | 4 | 523 | |||
U23765.1 | - | 4 | 387 | |||
X84213.1 | - | 4 | 515 |