Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001188.3 | NP_001179.1 | 6 | 914 | ||
XM_011514779.2 | XP_011513081.1 | 7 | 1025 | |||
XM_011514780.1 | XP_011513082.1 | 6 | 835 | |||
GenBank mRNA | AF520590.1 | - | 5 | 551 | ||
AK091807.1 | - | 6 | 779 | |||
AK292093.1 | - | 7 | 952 | |||
AK293547.1 | - | 6 | 844 | |||
BC004431.1 | - | 1 | 825 | 2819507 | ||
BC110337.1 | - | 7 | 844 | 5755200 | ||
CR457419.1 | - | 5 | 614 | |||
CR627020.1 | - | 1 | 6248 | |||
KU177934.1 | - | 5 | 614 | |||
KU177935.1 | - | 4 | 740 | |||
U16811.1 | - | 6 | 814 | |||
U23765.1 | - | 6 | 678 | |||
X84213.1 | - | 6 | 806 |