Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPRR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207015.1 | NP_001193944.1 | 5, 6 | 768 | ||
NM_001207016.1 | NP_001193945.1 | 3, 4 | 558 | |||
NM_002849.3 | NP_002840.2 | 6, 7 | 1424 | |||
NM_130846.2 | NP_570897.2 | 2, 3 | 690 | |||
XM_011538615.2 | XP_011536917.1 | 6, 7 | 1020 | |||
GenBank mRNA | AK295951.1 | - | 5, 6 | 758 | ||
AK304457.1 | - | 3, 4 | 517 | |||
AK304672.1 | - | 3, 4 | 558 | |||
AK312376.1 | - | 6, 7 | 1337 | |||
BC092469.1 | - | 2, 3 | 690 | |||
BC110900.1 | - | 2, 3 | 387 | 4538615 | ||
BX571751.1 | - | 2, 3 | 743 | |||
D64053.1 | - | 6, 7 | 1426 | |||
U42361.1 | - | 6, 7 | 1184 | |||
U77916.1 | - | 3, 4 | 408 | |||
U77917.1 | - | 2, 3 | 667 | |||
X82635.1 | - | 2, 3 | 648 | |||
Z79693.1 | - | 6, 7 | 1095 |