Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002853.3 | NP_002844.1 | 6 | 1073 | ||
NR_026591.1 | 5 | 915 | ||||
GenBank mRNA | AF011905.1 | - | 6 | 958 | ||
AF030933.1 | - | 8 | 1181 | |||
AF058392.1 | - | 5 | 744 | |||
AF058393.1 | - | 4 | 635 | |||
AF073524.1 | - | 6 | 939 | |||
AF074717.1 | - | 6 | 883 | |||
AF076841.1 | - | 6 | 939 | |||
AF084512.1 | - | 6 | 948 | |||
AF084513.1 | - | 5 | 839 | |||
AF090170.1 | - | 5 | 830 | |||
AJ004974.1 | - | 6 | 907 | |||
AJ004975.1 | - | 5 | 798 | |||
AK002112.1 | - | 6 | 1076 | |||
BC006837.1 | - | 6 | 925 | 3450321 | ||
BC009804.1 | - | 6 | 900 | 4109436 | ||
BC037857.1 | - | 6 | 2172 | 5272408 | ||
BT006908.1 | - | 5 | 744 |