Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BBS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024649.4 | NP_078925.3 | 11, 12 | 1130 | ||
GenBank mRNA | AF503941.1 | - | 11, 12 | 1130 | ||
AK095638.1 | - | 10, 11 | 1137 | |||
AK294962.1 | - | 3, 4 | 364 | |||
AK296311.1 | - | 7, 8 | 845 | |||
BC047642.1 | - | 10, 11 | 1089 | 4824649 | ||
BC109064.2 | - | 11, 12 | 1123 | 40033833 | ||
BX647612.1 | - | 11, 12 | 1159 |