Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCYT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184917.2 | NP_001171846.1 | 13 | 1183 | ||
NM_001256433.2 | NP_001243362.1 | 11 | 967 | |||
NM_001256434.2 | NP_001243363.1 | 13 | 1181 | |||
NM_001256435.2 | NP_001243364.1 | 12 | 1413 | |||
NM_001282203.1 | NP_001269132.1 | 12 | 1541 | |||
NM_001282204.1 | NP_001269133.1 | 12 | 1257 | |||
NM_002861.4 | NP_002852.1 | 12 | 1129 | |||
XM_005256386.3 | XP_005256443.1 | 13 | 1333 | |||
XM_005256387.3 | XP_005256444.1 | 13 | 1253 | |||
XM_006722287.3 | XP_006722350.1 | 12 | 1042 | |||
XM_017024910.1 | XP_016880399.1 | 11 | 988 | |||
XM_017024911.1 | XP_016880400.1 | 12 | 1199 | |||
XM_017024912.1 | XP_016880401.1 | 13 | 1305 | |||
GenBank mRNA | AK093328.1 | - | 12 | 1477 | ||
AK297996.1 | - | 12 | 1190 | |||
AK301723.1 | - | 12 | 1291 | |||
AK315777.1 | - | 13 | 1105 | |||
AK316385.1 | - | 12 | 950 | |||
AY251539.1 | - | 13 | 1074 | |||
BC000351.2 | - | 12 | 1037 | 2961418 | ||
BC010075.2 | - | 12 | 1080 | 4303545 | ||
BX419403.2 | - | 11 | 907 | |||
CR456779.1 | - | 12 | 1007 | |||
D84307.1 | - | 12 | 1073 |